Learn More
FSD1 Monoclonal Antibody (1G3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA522569
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MPDEDSKIDH YVLEYRRTNF EGPPRLKEDQ PWMVIEGIRQ TEYTLTGLKF DMKYMNFRVK ACNKAVAGEF SEPVTLETPA FMFRLDASTS HQNLRVDDLS VEWDAMGGKV QDIKAREKDG KGRTASPINS PARGTPSPKR MPSGRGGRDR FTAESYTVLG DTLIDGGEHY WEVRYEPDSK AFGVGVAYRS LGRFEQLGKT AASWCLHVNN WLQVSFTAKH ANKVKVLDAP VPDCLGVHCD FHQGLLSFYN ARTKQ
FSD1 Monoclonal antibody specifically detects FSD1 in Human samples. It is validated for ELISA, Immunoprecipitation.Specifications
FSD1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9BTV5 | |
FSD1 | |
FSD1 (-, 1 a.a. ∽ 309 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation | |
1G3 | |
Unconjugated | |
FSD1 | |
fibronectin type 3 and SPRY (spla, ryanodine) domain containing (with coiled-coil motif) 1; fibronectin type 3 and SPRY domain containing 1; fibronectin type 3 and SPRY domain-containing protein; fibronectin type III and SPRY domain containing 1; fibronectin type III and SPRY domain-containing protein 1; Fsd1; GLFND; Microtubule-associated protein GLFND; MID1-related protein 1; midline 1-related protein 1; MIR1; VLP27 | |
Mouse | |
Affinity chromatography | |
RUO | |
79187 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |