missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-8 Rat anti-Mouse, Clone: 163842, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB112-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438365

  • £193.88 / 50µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Frizzled-8 Monoclonal antibody MAB112SP Specifically detects Frizzled-8 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Frizzled-8
163842
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FZD8
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-8 Ala28-Pro173 (predicted) Accession # Q61091
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8325
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q61091
frizzled (Drosophila) homolog 8, frizzled 8, frizzled family receptor 8, frizzled homolog 8 (Drosophila), frizzled-8, Frizzled8, FZ-8, FZD8, hFz8
Rat
IgG2a
50 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Frizzled-8 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse (rm) Frizzled-6 is observed and no cross-reactivity with rmFrizzled-1, -2, -3, -4, or -7 is observed.
Mouse
Videos
Documents