missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-8 Rat anti-Mouse, Clone: 163842, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Frizzled-8 |
---|---|
Clone | 163842 |
Host Species | Rat |
Regulatory Status | RUO |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-8 Ala28-Pro173 (predicted) Accession # Q61091 |
Description
Frizzled-8 Monoclonal specifically detects Frizzled-8 in Mouse samples. It is validated for Western Blot.Specifications
Frizzled-8 | |
Rat | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-8 Ala28-Pro173 (predicted) Accession # Q61091 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
163842 | |
RUO | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant |