missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZD7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132790
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-32790 detects Frizzled 7 from human samples.
FZD7 Polyclonal antibody specifically detects FZD7 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FZD7 | |
Polyclonal | |
Unconjugated | |
FZD7 | |
frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O75084 | |
FZD7 | |
Synthetic 16 amino acid peptide from N-terminal extracellular domain of human Frizzled 7 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |