Learn More
Frizzled-2 Rat anti-Human, Mouse, Clone: 202102, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1307-050
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438535
  • £186.69 / 50µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Frizzled-2 Monoclonal antibody MAB1307SP Specifically detects Frizzled-2 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Frizzled-2
202102
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FZD2
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-2 Gln29-Leu168 Accession # Q9JIP6
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2535
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation
Q9JIP6
frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Frizzled2, fz-2, Fz2, FZD2, FzE2, hFz2
Rat
IgG2a
50 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Frizzled-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Frizzled-1, -3, -4, -6, -7, -8, or rhFrizzled-5.
Human, Mouse