Learn More
FZD10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527174
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: H1299, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FZD10 Polyclonal antibody specifically detects FZD10 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FZD10 | |
Polyclonal | |
Unconjugated | |
FZD10 | |
CD350; CD350 antigen; frizzled 10, seven transmembrane spanning receptor; frizzled class receptor 10; frizzled family receptor 10; frizzled homolog 10; frizzled homolog 10 (Drosophila); frizzled homolog 10, pseudogene 1; frizzled-10; Fz10; Fz-10; Fzd10; Fzd10-ps1; FzE7; hFz10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11211, 93897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7 | |
Q8BKG4, Q9ULW2 | |
FZD10 | |
Synthetic peptide corresponding to a region within amino acids 121 and 581 of Human Frizzled 10 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |