Learn More
FZD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528757
Code : PN
Additional Details : Weight : 0.01000kg
Description
PA5-28757 targets Frizzled-1 in WB applications and shows reactivity with Human samples. The PA5-28757 immunogen is a synthetic peptide corresponding to a region within amino acids 1 and 45 of Frizzled 1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.Specifications
FZD1 | |
Polyclonal | |
Unconjugated | |
Q9UP38 | |
FZD1 | |
Synthetic peptide corresponding to a region within amino acids 1 and 45 of Frizzled 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.22 mg/mL | |
FZD1 | |
Wnt receptor, frizzled 1, seven transmembrane spanning receptor, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, fz-1, fzE1, hFz1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8321 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |