Learn More
Frizzled-1 Rat anti-Mouse, Clone: 162526, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1120-500
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448365
  • £376.32 / 500µg
Estimated Shipment date
from Supplier 19-11-2019
Description
Description
Frizzled-1 Monoclonal antibody MAB1120500 Specifically detects Frizzled-1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Frizzled-1
162526
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FZD1
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1 Gln72-His248 (Met122Ile) Accession # O70421
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8321
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O70421
DKFZp564G072, FLJ95923, frizzled family receptor 1, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, Frizzled1, fz-1, FZD1, fzE1, hFz1, seven transmembrane spanning receptor, Wnt receptor
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Frizzled-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-2, -3, -4, -6, -7, or -8 is observed.