missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533534
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Monkey (100%) Gorilla (94%) Orangutan (88%).
FPR1 Polyclonal antibody specifically detects FPR1 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FPR1 | |
Polyclonal | |
Unconjugated | |
FPR1 | |
fMet-Leu-Phe receptor; fMLF-R; FMLP; fMLP receptor; formyl peptide receptor 1; FPR; Fpr receptor; FPR1; FPR-1; lipoxin A4 receptor; LXA4R; N-formyl peptide receptor; N-formylpeptide chemoattractant receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P21462 | |
FPR1 | |
Synthetic 16 amino acid peptide from 3rd extracellular domain of human FPR1. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |