Learn More
FPR1 Mouse anti-Human, Clone: 350418, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3744
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450236
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
FPR1 Monoclonal antibody MAB3744 Specifically detects FPR1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
FPR1
350418
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FPR1
HEK293 human embryonic kidney cell line transfected with human FPR1 Met1-Lys350 Accession # NP_002020
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2357
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
NP_002020
fMet-Leu-Phe receptor, fMLF-R, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human FPR1. Stains human FPR1-transfected cells but not irrelevant transfectants.