missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXRED2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534345
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human lung tissue lysate. PA5-34345 can be used with blocking peptide PEP-1388.
FOXRED2 Polyclonal antibody specifically detects FOXRED2 in Human samples. It is validated for Western Blot.Specifications
FOXRED2 | |
Polyclonal | |
Unconjugated | |
FOXRED2 | |
Endoplasmic reticulum flavoprotein associated with degradation; ER flavoprotein associated with degradation; ERFAD; FAD dependent oxidoreductase domain containing 2; FAD-dependent oxidoreductase domain containing 2; FAD-dependent oxidoreductase domain-containing protein 2; FOXRED2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80020 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8IWF2 | |
FOXRED2 | |
A 17 amino acid synthetic peptide near the amino terminus of human FOXRED2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |