Learn More
FOXP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530908
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, HeLa nucleus, mouse brain. Predicted reactivity: Mouse (94%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FOXP4 Polyclonal antibody specifically detects FOXP4 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
FOXP4 | |
Polyclonal | |
Unconjugated | |
Foxp4 | |
1200010K03Rik; 2310007G05Rik; FKHLA; FLJ40908; FLJ44184; fork head-related protein like A; fork head-related protein-like A; forkhead box P4; forkhead box protein P4; Foxp4; hFKHLA; mFKHLA; OTTHUMP00000043536; winged-helix repressor FOXP4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116113, 74123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.73 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IVH2, Q9DBY0 | |
Foxp4 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 275 of Human FOXP4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |