Learn More
FOXP3 Recombinant Rabbit Monoclonal Antibody (5H10L18), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 700914
Code : NEW
Additional Details : Weight : 1.00000kg
Description
This antibody is predicted to react with bovine, feline, porcine and rat based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
FOXP3 Recombinant Monoclonal antibody specifically detects FOXP3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FOXP3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
proprietary | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
5H10L18 | |
Unconjugated | |
Q9BZS1 | |
Rabbit | |
Protein A | |
RUO | |
50943 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |