missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXP3 Monoclonal Antibody (3G3), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518160
Code : NEW
Additional Details : Weight : 0.01000kg
Description
FOXP3 Monoclonal antibody specifically detects FOXP3 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
FOXP3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q99JB6, Q9BZS1 | |
Foxp3 | |
Full-length His-tagged recombinant murine FoxP3 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Western Blot | |
3G3 | |
Alexa Fluor 647 | |
Foxp3 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
20371, 50943 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |