Learn More
FOXL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540518
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: HLFDPRLPAL AASPMLYLYG PERPGLPLAF APAAALAASG RAETPQKPPY Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 92%; Rat: 86%
FOXL1 Polyclonal antibody specifically detects FOXL1 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FOXL1 | |
Polyclonal | |
Unconjugated | |
FOXL1 | |
FKH11; Fkh6; fkh-6; FKHL10; FKHL11; forkhead box L1; forkhead box protein L1; forkhead homolog 6; Forkhead related transcription factor; forkhead-like 11; forkhead-related protein FKHL11; Forkhead-related transcription factor 7; Foxl1; FREAC6; FREAC7; FREAC-7; JFJ-3; Transcription factor FKH-6 | |
Rabbit | |
Protein A | |
RUO | |
14241, 2300 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q12952, Q64731 | |
FOXL1 | |
Synthetic peptide directed towards the N-terminal of human FOXL1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |