missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535097
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
FOXA2 Polyclonal antibody specifically detects FOXA2 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FOXA2 | |
Polyclonal | |
Unconjugated | |
FOXA2 | |
FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
25099, 3170 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P32182, Q9Y261 | |
FOXA2 | |
KLH conjugated synthetic peptide between 380-407 amino acids from the C-terminal region of human FOXA2 | |
400 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |