Learn More
Follistatin-related Gene Protein/FLRG Rat anti-Mouse, Clone: 210725, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1255
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448495
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Follistatin-related Gene Protein/FLRG Monoclonal antibody MAB1255 Specifically detects Follistatin-related Gene Protein/FLRG in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Follistatin-related Gene Protein/FLRG/Fstl3
210725
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FSTL3
Mouse myeloma cell line NS0-derived recombinant mouse FLRG Val24-Val256 Accession # Q9EQC7
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10272
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9EQC7
FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse FLRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 50-100% cross-reactivity with recombinant human FLRG.