missing translation for 'onlineSavingsMsg'
Learn More

Follistatin-like 1/FSTL1 Rat anti-Mouse, Clone: 228210, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1738

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428875

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Follistatin-like 1/FSTL1 Monoclonal antibody MAB1738 Specifically detects Follistatin-like 1/FSTL1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Follistatin-like 1/FSTL1
228210
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FSTL1
Mouse myeloma cell line NS0-derived recombinant mouse Follistatin-like 1/FSTL1 Glu19-Ile306 Accession # Q62356
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
11167
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q62356
Flik, FLJ50214, follistatin-like 1, Follistatin-like protein 1, Follistatin-related Protein, follistatin-related protein 1, FRP, FRPFLJ52277, FSL1, FSTL1, TSC-36
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Follistatin-like 1/FSTL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human Follistatin-like 1/FSTL1 is observed.
Mouse
Videos
Documents

For Research Use Only