Learn More
DYKDDDDK Tag Monoclonal Antibody (L5), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA11421MG
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The MA1-142 antibody (clone L5) has been successfully used as a detection antibody in a direct ELISA with FLAG-tagged protein-coated plates. Similar Epitope as the FLAG tag from Sigma. The MA1-142 immunogen is recombinant fusion proteins of mouse Langerin/CD207, which contained a flexible linker sequence from E. coli OmpF and a FLAG epitope. (J Immunol Methods. 2008 Feb 29;331(1-2):27-38. Epub 2007 Nov 26.)
DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for ELISA, Western Blot.Specifications
DYKDDDDK Tag | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rat | |
Protein G | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
ELISA, Western Blot | |
L5 | |
Unconjugated | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
Recombinant fusion proteins of mouse Langerin/CD207 | |
1 mg | |
Primary | |
Antibody | |
IgG2a |