missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-2 Mouse anti-Human, Clone: 297018, R&D Systems™
Mouse Monoclonal Antibody
Brand: RD Systems (Bio-Techne) MAB2428
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Ficolin-2 Monoclonal antibody MAB2428 Specifically detects Ficolin-2 in Human samples. It is validated for Western BlottingSpecifications
Ficolin-2 | |
297018 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Purified | |
FCN2 | |
Mouse myeloma cell line NS0-derived recombinant human Ficolin-2 Leu26-Ala313 Accession # Q15485 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
2220 | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q15485 | |
37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCN2, FCNL, FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin2, Ficolin-B, Ficolin-beta, Hucolin, L-Ficolin, P35, Serum lectin p35 | |
Mouse | |
IgG2b | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Monoclonal | |
Detects human Ficolin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Ficolin-3 is observed. | |
Human |
For Research Use Only