Learn More
Fibrinogen Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA185429
Code : LD
Additional Details : Weight : 0.01000kg
Description
Sheep anti Human fibrinogen antibody recognizes human fibrinogen, a complex approximately 340 kDa hetero-hexameric (di-trimeric) glycoprotein consisting of 3 pairs of alpha, beta, and gamma chains linked by a series of 29 disulphide bonds (Henschen et al. 1983).
Fibrinogen Polyclonal antibody specifically detects Fibrinogen in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen).Specifications
Fibrinogen | |
Polyclonal | |
Unconjugated | |
FGG | |
2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; carboxy-terminal region; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; Fgb; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen alpha-E subunit; fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma polypeptide; fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
Sheep | |
Ion-exchange chromatography | |
RUO | |
110135, 14161, 2243, 2244, 2266, 24366, 24367, 361969, 99571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen) | |
5 mg/mL | |
glycine buffered saline with <0.01% benzamidine, <0.1% EACA, <1mM EDTA and <0.1% sodium azide; pH 7.4 | |
E9PV24, P02671, P02675, P02679, P02680, P06399, P14480, Q8K0E8, Q8VCM7 | |
FGA, FGB, FGG | |
Human fibrinogen purified from plasma. | |
1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |