missing translation for 'onlineSavingsMsg'
Learn More

FHL2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA112859

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11500523

  • £377.79 / 50µg


Estimated Shipment
22-06-2018
Add to basket

Description

Description

A suggested positive control for this product is Hela cell lysate.
Specifications

Specifications

FHL2
Polyclonal
PBS with 0.08% sodium azide
Q14192
DRAL, SLIM3
Rabbit
IgG
50μg
Antibody
Polyclonal
Human
Western Blot
Unconjugated
FHL2
Purified
FHL2
Synthetic peptide corresponding to human FHL2.
Antigen affinity chromatography
Store at -80°C, Avoid Freeze/Thaw Cycles
Primary
2274
Videos