missing translation for 'onlineSavingsMsg'
Learn More

FHL2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA112870

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11510533

  • £260.94 / 100µg


Estimated Shipment
29-05-2018
Add to basket

Description

Description

A suggested positive control for this product is HeLa cell lysate.
Specifications

Specifications

FHL2
Polyclonal
PBS with 0.08% sodium azide
Q14192
AAG11, DRAL, SLIM3
Rabbit
IgG
100μg
Antibody
Polyclonal
Human
Western Blot
Unconjugated
FHL2
Purified
FHL2
Synthetic peptide corresponding to human FHL2.
Ammonium sulfate precipitation
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
2274
Videos