Learn More
FGF-9 Mouse anti-Human, Clone: 36923, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2731
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459655
  • £337.93 / 500µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
FGF-9 Monoclonal antibody MAB2731 Specifically detects FGF-9 in Human samples. It is validated for Western Blotting
Specifications
Specifications
FGF-9
36923
Western Blot 1 ug/mL
P31371
FGF9, FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human FGF-9 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF-3, -4, -5, -7, -10, -11, -13, -16, -17, -18, -19, rhFGF acidic, rhFGF basic, recombinant mouse (rm) FGF-6, -8b, -8c, or -15 is observed. In Western blots, no cross-reactivity with rhFGF-3, -4, -5, -6, -7, -10, -13, rmFGF-8b, or rmFGF-8c is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FGF9
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF-9 Met1-Ser208 Accession # P31371
Protein A or G purified from ascites
RUO
Primary
2254
Reconstitute at 0.5 mg/mL in sterile PBS.