missing translation for 'onlineSavingsMsg'
Learn More

FGF-20 Rat anti-Human, Clone: 272317, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2547

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459525

  • £279.85 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

FGF-20 Monoclonal antibody MAB2547 Specifically detects FGF-20 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

FGF-20
272317
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FGF20
E. coli-derived recombinant human FGF-20, Met1-Thr211, Accession # Q9NP95
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
26281
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Q9NP95
FGF20, FGF-20, fibroblast growth factor 20
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human FGF-20 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) FGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -16, -17, -18, -19, -23, recombinant mouse FGF-8b, -8c, -15, -23, rhFGF acidic, or rhFGF basic is observed.
Human
Videos

For Research Use Only