missing translation for 'onlineSavingsMsg'
Learn More

FGF-16 Mouse anti-Human, Clone: 209511, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1212

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438455

  • £337.93 / 500µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FGF-16 Monoclonal antibody MAB1212 Specifically detects FGF-16 in Human samples. It is validated for Western Blotting
Specifications

Specifications

FGF-16
209511
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FGF16
E. coli-derived recombinant human FGF-16 Ala2-Arg207 Accession # O43320
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8823
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O43320
FGF16, FGF-16, fibroblast growth factor 16
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human FGF-16 in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant human (rh) FGF-3, -5, -8c, and -11 is observed and no cross-reactivity with rhFGF acidic, rhFGF basic, rhFGF-4, -6, -7, -8, -9, -10, -12, -13, -17, -18, or -19 is observed.
Human
Videos

For Research Use Only