Learn More
FGF-11 Mouse anti-Human, Clone: 224812, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1634
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488785
  • £358.00 / 500µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
FGF-11 Monoclonal antibody MAB1634 Specifically detects FGF-11 in Human samples. It is validated for Western Blotting
Specifications
Specifications
FGF-11
Unconjugated
Murine
IgG2b
500μg
Primary
Human
224812
Purified
E.Coli-derived recombinant human FGF-11. Met1-Pro225 (predicted)
Protein A or G purified from hybridoma culture supernatant