Learn More
Fetuin A/AHSG Rat anti-Mouse, Clone: 207440, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB15632
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438735
  • £376.32 / 500µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Fetuin A/AHSG Monoclonal antibody MAB15632 Specifically detects Fetuin A/AHSG in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
Fetuin A/AHSG
207440
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
AHSG
Mouse myeloma cell line NS0-derived recombinant mouse Fetuin A/AHSG Ala19-Ile345 Accession # P29699
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
197
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P29699
A2HS, AHS, AHSG, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Fetuin A/AHSG in ELISAs. In sandwich immunoassays, no cross-reactivty or interference was observed with recombinant human Fetuin A, rhTGF-beta 1, recombinant mouse (rm) BMP-4, rmMMP-2, recombinant rat MMP-2, or rmMMP-9 is observed.