missing translation for 'onlineSavingsMsg'
Learn More

IL-10 Goat anti-Feline, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF736

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13450095

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-10 Polyclonal antibody AF736 Specifically detects IL-10 in Feline samples. It is validated for Western Blotting
Specifications

Specifications

IL-10
LYOPH
Western Blot 0.1 ug/mL
P55029.2
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects feline IL-10 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant canine IL-10, recombinant human IL-10, and recombinant porcine IL-10 is observed and 10% cross-reactivity with recombinant mouse IL-10 and recombinant rat IL-10 is observed.
Feline
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL10
E. coli-derived recombinant feline IL-10 Ser19-Ile178 Accession # P55029.2
Immunogen affinity purified
RUO
Primary
3586
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only