missing translation for 'onlineSavingsMsg'
Learn More

CD4 Goat anti-Feline, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2597

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437664

  • £421.89 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD4 Polyclonal antibody AF2597 Specifically detects CD4 in Feline samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blotting
Specifications

Specifications

CD4
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Affinity Purified
CD4
Mouse myeloma cell line NS0-derived recombinant feline CD4
Antigen Affinity-purified
RUO
Primary
920
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects feline CD4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant canine CD4 is observed and 5% cross-reactivity with recombinant human CD4 and recombinant mouse CD4 is observed.
Feline
Videos
Documents

For Research Use Only