missing translation for 'onlineSavingsMsg'
Learn More

FCRLB/FCRY Rat anti-Mouse, Clone: 465905, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4868

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460746

  • £249.49 / 100µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

FCRLB/FCRY Monoclonal antibody MAB4868 Specifically detects FCRLB/FCRY in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

FCRLB/FCRY
465905
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FCRLB
Mouse myeloma cell line NS0-derived recombinant mouse FCRLB/FCRY Ala18-Ser427 Accession # Q5DRQ8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
127943
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q5DRQ8
Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FcRL2, FCRL2FCRLM2, FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2, FREB-2FcRY, RP11-474I16.6
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse FCRLB/FCRY in direct ELISAs and Western blots. In Western blots, 75% cross-reactivity with recombinant human FCRLB is observed.
Mouse
Videos

For Research Use Only