missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCAMR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545118
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ATLSQTPAVG PWGPPGKESS VKRTFPEDES SSRTLAPVST MLALFMLMAL Sequence homology: Human: 100%
FCAMR Polyclonal antibody specifically detects FCAMR in Human samples. It is validated for Western Blot.Specifications
FCAMR | |
Polyclonal | |
Unconjugated | |
FCAMR | |
CD351; fc alpha/mu receptor; Fc fragment of IgA and IgM receptor; Fc IgA/IgM receptor; Fc receptor, IgA, IgM, high affinity; Fca/m receptor; FCA/MR; Fcamr; FKSG87; high affinity immunoglobulin alpha and immunoglobulin mu Fc receptor; immunity related factor; mFcamR; receptor for Fc fragment of IgA and IgM | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83953 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WWV6 | |
FCAMR | |
Synthetic peptide directed towards the C-terminal of human FCAMR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |