Learn More
FBXO4 Monoclonal Antibody (2F2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA521639
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MAGSEPRSGT NSPPPPFSDW GRLEAAILSG WKTFWQSVSK ERVARTTSRE EVDEAASTLT RLPIDVQLYI LSFLSPHDLC QLGSTNHYWN ETVRDPILWR YFLLRDLPSW SSVDWKSLPD LEILKKPISE VTDGAFFDYM AVYRMCCPYT RRASKSSRPM YGAVTSFLHS LIIQNEPRFA MFGPGLEELN TSLVLSLMSS EELCPTAGLP QRQIDGIGSG VNFQLNNQHK FNILILYSTT RKERDRAREE HTSAVNKMFS RHNEGDDQQG SRYSVIPQIQ KVCEVVDGFI YVANAEAHKR HEWQDEFSHI MAMTDPAFGS SGRPLLVLSC ISQGDVKRMP CFYLAHELHL NLLNHPWLVQ DTEAETLTGF LNGIEWILEE VESKRAR
FBXO4 Monoclonal antibody specifically detects FBXO4 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
FBXO4 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9UKT5 | |
FBXO4 | |
FBXO4 (AAH48098, 1 a.a. ∽ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
2F2 | |
Unconjugated | |
FBXO4 | |
1700096C12Rik; AI851261; AW494535; F-box only protein 4; F-box protein 4; F-box protein Fbx4; FBX4; Fbxo4 | |
Mouse | |
Affinity chromatography | |
RUO | |
26272 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |