missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531380
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa. Predicted reactivity: Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FBXL8 Polyclonal antibody specifically detects FBXL8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FBXL8 | |
Polyclonal | |
Unconjugated | |
FBXL8 | |
FBL8; F-box and leucine rich repeat protein 8; F-box and leucine-rich repeat protein 8; F-box protein FBL8; F-box/LRR-repeat protein 8; FBXL8; RGD1566420 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96CD0 | |
FBXL8 | |
Recombinant fragment corresponding to a region within amino acids 1 and 205 of Human FBL8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |