Learn More
FBXL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530510
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, human FBXL4-transfected 293T. Predicted reactivity: Mouse (95%), Pig (94%), Rhesus Monkey (98%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FBXL4 Polyclonal antibody specifically detects FBXL4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FBXL4 | |
Polyclonal | |
Unconjugated | |
FBXL4 | |
AI836810; FBL4; FBL5; F-box and leucine rich repeat protein 4; F-box and leucine-rich repeat protein 4; F-box protein FBL4/FBL5; F-box/LRR-repeat protein 4; Fbxl4; MTDPS13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UKA2 | |
FBXL4 | |
Recombinant fragment corresponding to a region within amino acids 366 and 608 of Human FBL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |