missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBG4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA123512
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-23512 has been successfully used in ELISA and Western Blot procedures.
FBG4 Polyclonal antibody specifically detects FBG4 in Human samples. It is validated for ELISA, Western Blot.Specifications
FBG4 | |
Polyclonal | |
Unconjugated | |
FBXO17 | |
F Box Gama 4; FBG4; F-box only protein 17; F-box only protein 26; F-box protein 17; F-box protein FBG4; FBX17; FBX26; Fbxo17; FBXO26; FLJ11798; FLJ25205; FLJ58300; MGC9379 | |
Rabbit | |
Affinity chromatography | |
RUO | |
115290 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q96EF6 | |
FBXO17 | |
Rabbit Polyclonal to FBG4 ( F Box 4 ) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |