missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC27A4 Monoclonal Antibody (342142), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524075
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This antibody stains human FATP4-transfected cells but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
SLC27A4 Monoclonal antibody specifically detects SLC27A4 in Human samples. It is validated for Flow Cytometry.Specifications
SLC27A4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6P1M0 | |
SLC27A4 | |
NS0 mouse myeloma cell line transfected withhuman FATP4 Met1-Leu643 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
342142 | |
Unconjugated | |
SLC27A4 | |
ACSVL4; BB144259; Fatp4; FATP-4; fatty acid transport protein 4; IPS; Long-chain fatty acid transport protein 4; SLC27A4; solute carrier family 27 (fatty acid transporter), member 4; solute carrier family 27 member 4 | |
Mouse | |
Protein A/G | |
RUO | |
10999 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |