missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas Ligand/TNFSF6 Mouse anti-Rat, Clone: 252029, R&D Systems™
Mouse Monoclonal Antibody
Brand: RD Systems (Bio-Techne) MAB1858
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Fas Ligand/TNFSF6 Monoclonal antibody MAB1858 Specifically detects Fas Ligand/TNFSF6 in Rat samples. It is validated for Immunocytochemistry, Western BlottingSpecifications
Fas Ligand/TNFSF6 | |
252029 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Purified | |
FASLG | |
Mouse myeloma cell line NS0-derived recombinant rat Fas Ligand/TNFSF6 Leu104-Leu278 Accession # P36940 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
356 | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P36940 | |
apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
Mouse | |
IgG2b | |
500 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Monoclonal | |
Detects rat Fas Ligand/TNFSF6 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) 4-1BB Ligand, recombinant human (rh) APRIL, rmBAFF, rhEDA-A2, rmEDA, rhFas Ligand, rmFas Ligand, rhGITR Ligand, rmLIGHT, rhLymphotoxin alpha 1/beta 2, rhLymphotoxin alpha 2/beta 1, rmOX40Ligand, rhTL1, recombinant cotton rat TNF-alpha , rmTNF-alpha , rrTNF-alpha , rmTrail, rhTrance, and rmTWEAK is observed. | |
Rat |
For Research Use Only