Learn More
FAS Monoclonal Antibody (GH-9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17624
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-7624 detects FAS from human, rat and mouse samples. MA1-7624 has been successfully used in ELISA, Western blot and immunoprecipitation procedures. The MA1-7624 immunogen is a purified recombinant FAS protein.
FAS Monoclonal antibody specifically detects FAS in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
FAS | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
P25445, P25446, Q63199 | |
FAS | |
Purified recombinant FAS protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Western Blot | |
GH-9 | |
Unconjugated | |
FAS | |
AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; Apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; FAS; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 26; Tumor necrosis factor receptor superfamily, member 6 | |
Mouse | |
Protein A | |
RUO | |
14102, 246097, 355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |