missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518620
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
FXR Polyclonal antibody specifically detects FXR in Human samples. It is validated for Western Blot.Specifications
FXR | |
Polyclonal | |
Unconjugated | |
NR1H4 | |
AI957360; BAR; bile acid receptor; farnesoid X activated receptor; farnesoid X nuclear receptor; farnesoid X-activated receptor; Farnesol receptor HRR-1; FXR; HRR1; HRR-1; hypothetical protein LOC540528; MGC163445; NR1H4; Nuclear receptor subfamily 1 group H member 4; nuclear receptor subfamily 1, group H, member 4; retinoid X receptor-interacting protein 14; RIP14; RXR-interacting protein 14; Rxrip14 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9971 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96RI1 | |
NR1H4 | |
Synthetic peptide sequence (KSCREKTELTPDQQ) corresponding to the internal amino acids of NR1H4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |