missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCB Monoclonal Antibody (2B10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518862
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: GSENFLIDNM AFTLEKELVT LSSLSSAIAK HESNFMQRCE VSKGKSSVVA AALSDRRENI HPYRKELQRE KKKMLQTNLK VSGALYREIT LKVAEVQLKS DFAAQKLSN
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group B. Alternative splicing results in two transcript variants encoding the same protein.Specifications
FANCB | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q8NB91 | |
Mouse | |
Affinity chromatography | |
RUO | |
2187 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2B10 | |
Unconjugated | |
FANCB | |
FANCB | |
FANCB (NP_689846.1, 750 a.a. ∽ 858 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |