missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146122
Code : LD
Additional Details : Weight : 0.01000kg
Description
FANCA Polyclonal antibody specifically detects FANCA in Human samples. It is validated for Western Blot.Specifications
FANCA | |
Polyclonal | |
Unconjugated | |
FANCA | |
AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
O15360 | |
FANCA | |
Synthetic peptide within residues 1200-1300 of the human protein. | |
RUO | |
2175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |