missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM82A1 Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Thermo Scientific PA520938
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is K562 cell lysate. PA5-20938 can be used with blocking peptide PEP-1052.
FAM82A1 Polyclonal antibody specifically detects FAM82A1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
FAM82A1 | |
Polyclonal | |
Unconjugated | |
RMDN2 | |
AW061290; BLOCK18; Fam82a; Fam82a1; family with sequence similarity 82, member A; family with sequence similarity 82, member A1; microtubule-associated protein; mRMD-2; PRO34163; Protein FAM82A1; PYST9371; regulator of microtubule dynamics 2; regulator of microtubule dynamics protein 2; RMD2; RMD-2; RMD4; Rmdn2; UNQ9371/PRO34163 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
151393, 313840, 381110 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q498D5, Q8BSE0, Q96LZ7 | |
RMDN2 | |
A 16 amino acid peptide near the amino terminus of human FAM82A1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |