Learn More
FAM3C Monoclonal Antibody (438618), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524162
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) FAM3A, rhFAM3B, recombinant mouse (rm) FAM3C, or rmFAM3D is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
FAM3C Monoclonal antibody specifically detects FAM3C in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
FAM3C | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q92520 | |
FAM3C | |
Mouse myeloma cell line NS0-derived recombinant human FAM3C Gln25-Asp227 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
438618 | |
Unconjugated | |
FAM3C | |
D6Wsu176e; FAM3 metabolism regulating signaling molecule C; Fam3c; fam3c protein; FAM3C-like protein; family with sequence similarity 3 member C; family with sequence similarity 3, member C; GS3786; Ilei; Interleukin-like EMT inducer; interleukin-like epithelial-mesenchymal transition inducer; predicted osteoblast protein; protein FAM3C; si:dkey-159a18.2; wu:fb99h11; wu:fi35h03; zgc:55444; zgc:77090 | |
Mouse | |
Protein A/G | |
RUO | |
10447 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |