Learn More
FAM3C Rat anti-Mouse, Clone: 372018, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2868
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469745
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
FAM3C Monoclonal antibody MAB2868 Specifically detects FAM3C in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
FAM3C
372018
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FAM3C
Mouse myeloma cell line NS0-derived recombinant mouse FAM3C Gln25-Asp227 Accession # Q91VU0
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10447
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q91VU0
D6WSU176e, family with sequence similarity 3, member C, GS3876, ILEI, ILEIInterleukin-like EMT inducer, predicted osteoblast protein
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse FAM3C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse FAM3A, 3B, or 3D is observed.