missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM174B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542240
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MRAVPLPAPL LPLLLLALLA APAARASRAE SVSAPWPEPE RESRPPPGPG Sequence homology: Human: 93%
FAM174B Polyclonal antibody specifically detects FAM174B in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FAM174B | |
Polyclonal | |
Unconjugated | |
FAM174B | |
fam174b; family with sequence similarity 174 member B; family with sequence similarity 174, member B; membrane protein FAM174B; si:ch211-196l7.3; Unknown (protein for MGC:140090) | |
Rabbit | |
Protein A | |
RUO | |
400451 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3ZCQ3 | |
FAM174B | |
Synthetic peptide directed towards the N-terminal of human LOC400451 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |