Learn More
FAM105A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543349
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: HKLKWWIGYL QRKFKRNLSV EAEVDLLSYC AREWKGETPR NKLMRKAYEE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
FAM105A Polyclonal antibody specifically detects FAM105A in Human samples. It is validated for Western Blot.Specifications
FAM105A | |
Polyclonal | |
Unconjugated | |
FAM105A | |
9830126M18; FAM105A; family with sequence similarity 105 member A; family with sequence similarity 105, member A; inactive ubiquitin thioesterase FAM105A; Inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase FAM105A; LOW QUALITY PROTEIN: inactive ubiquitin thioesterase FAM105A; NET20; OTU deubiquitinase with linear linkage specificity like; OTULINL; protein FAM105A; Protein FAM105A-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54491 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NUU6 | |
FAM105A | |
Synthetic peptide directed towards the N-terminal of human FAM105A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |