missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor X/Xa Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA143047
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-43047 detects Factor X in human samples. PA1-43047 has successfully been used in Western blot and ELISA applications. PA1-43047 was derived from human factor X.
Factor X/Xa Polyclonal antibody specifically detects Factor X/Xa in Human samples. It is validated for ELISA, Western Blot.Specifications
Factor X/Xa | |
Polyclonal | |
Unconjugated | |
F10 | |
Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product | |
Sheep | |
Ammonium sulfate precipitation, Ion-exchange chromatography | |
RUO | |
2159 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
15.2 mg/mL | |
50% water with 50% glycerol and no preservative | |
P00742 | |
F10 | |
Human Factor X. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |