Learn More
Factor VIII Monoclonal Antibody (F8-2.2.32), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110588
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10588 detects full-length human Factor VIII. This antibody does not cross-react with von Willebrand factor. MA1-10588 has been successfully used in ELISA and Western blot applications. The MA1-10588 immunogen is purified human Factor VIII.
Factor VIII Monoclonal antibody specifically detects Factor VIII in Human samples. It is validated for ELISA, Western Blot.Specifications
Factor VIII | |
Monoclonal | |
0.72 mg/mL | |
PBS with no preservative; pH 7.4 | |
P00451 | |
F8 | |
Purified human Factor VIII. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
F8-2.2.32 | |
Unconjugated | |
F8 | |
AHF; Antihemophilic factor; CF8; Cf-8; coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIII, procoagulant component (hemophilia A); coagulation factor VIIIc; DXS1253E; F8; F8B; F8C; Factor 8; Factor VIII; Factor VIII B chain; factor VIII F8B; factor VIII precursor; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; factor8; FVIII; HEMA; Procoagulant component; RP11-115M6.7 | |
Mouse | |
Protein A | |
RUO | |
2157 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |