missing translation for 'onlineSavingsMsg'
Learn More

anti-Factor IX Polyclonal, Invitrogen™

Factor IX Polyclonal antibody PA1-74023 from Thermo Scientific™ specifically detects Factor IX in Human samples, and it is validated for ELISA and Immunoelectrophoresis

Brand:  Thermo Scientific Pierce PA1-74023

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11548591

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description


Host Species: Goat
Cross Reactivity: Human
Isotype: IgG
Immunogen: Factor IX from human plasma.
Formulation: 200μg purified Antibody in proprietary buffer, with 50% glycerol and no preservative;
Uniprot Accession: P00740
Gene Symbol: F9
Entrez GeneID: 2158
PA1-74023 detects Factor IX in Human samples. PA1-74023 has been successfully used in ELISA and Immunoelectrophoresis procedures. PA1-74023 The immunogen is Factor IX from human plasma..
Specifications

Specifications

Factor IX
HRP
P00740
F9
Factor IX from human plasma.
200μg
Primary
2158
ELISAĆ¾Immunoelectrophoresis
200μg purified Antibody in proprietary buffer, with 50% glycerol and no preservative;
Purified
Goat
IgG
store at -20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Human
Videos